7-Bromo-5-methoxyquinoline ≥98%

Lieferant: Apollo Scientific

OR510163-100MG
APOSOR510163-100MGEA 59.6 EUR
APOSOR510163-100MG
7-Bromo-5-methoxyquinoline ≥98%
7-Bromo-5-methoxyquinoline
Formel: C₁₀H₈BrNO
Molecular Weight: 238,081 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11878498
CAS-Nummer: 1378860-76-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR