6-Bromo-4-methoxy-1H-indole-3-carbaldehyde ≥98%
Lieferant: Apollo Scientific
OR510166-100MG
APOSOR510166-100MGEA
150
EUR
APOSOR510166-100MG
6-Bromo-4-methoxy-1H-indole-3-carbaldehyde ≥98%
6-Bromo-4-methoxy-1H-indole-3-carbaldehyde
Formel:
C₁₀H₈BrNO₂ Molecular Weight: 254,08 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16610101 CAS-Nummer: 1202766-19-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...