7-Bromochromane-3-carboxylic acid ≥98%

Lieferant: Apollo Scientific

OR510167-100MG
APOSOR510167-100MGEA 369 EUR
APOSOR510167-100MG
7-Bromochromane-3-carboxylic acid ≥98%
7-Bromochromane-3-carboxylic acid
Formel: C₁₀H₉BrO₃
Molecular Weight: 257,081 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27922386
CAS-Nummer: 1781758-38-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR