1-Chloroisoquinoline-5-sulphonyl chloride ≥98%

Lieferant: Apollo Scientific

OR510168-100MG
APOSOR510168-100MGEA 253 EUR
APOSOR510168-100MG
1-Chloroisoquinoline-5-sulphonyl chloride ≥98%
1-Chloroisoquinoline-5-sulphonyl chloride
Formel: C₉H₅Cl₂NO₂S
Molecular Weight: 262,112 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11052630
CAS-Nummer: 141519-77-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR