(5-Bromo-1H-indazol-3-yl)methanamine hydrochloride ≥98%

Lieferant: Apollo Scientific

OR510169-100MG
APOSOR510169-100MGEA 417 EUR
APOSOR510169-100MG
(5-Bromo-1H-indazol-3-yl)methanamine hydrochloride ≥98%
(5-Bromo-1H-indazol-3-yl)methanamine hydrochloride
Formel: C₈H₈BrN₃·ClH
Molecular Weight: 262,534 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28893135
CAS-Nummer: 2436296-64-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR