2-Chloro-4-methoxy-5H-pyrrolo[3,2-d]pyrimidine ≥98%

Lieferant: Apollo Scientific

OR510171-100MG
APOSOR510171-100MGEA 449 EUR
APOSOR510171-100MG
2-Chloro-4-methoxy-5H-pyrrolo[3,2-d]pyrimidine ≥98%
2-Chloro-4-methoxy-5H-pyrrolo[3,2-d]pyrimidine
Formel: C₇H₆ClN₃O
Molecular Weight: 183,595 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21605181
CAS-Nummer: 1375301-68-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR