(7-Chloro-[1,2,4]triazolo[1,5-a]pyridin-2-yl)methanol ≥98%

Lieferant: Apollo Scientific

OR510172-100MG
APOSOR510172-100MGEA 0 EUR
APOSOR510172-100MG
(7-Chloro-[1,2,4]triazolo[1,5-a]pyridin-2-yl)methanol ≥98%
(7-Chloro-[1,2,4]triazolo[1,5-a]pyridin-2-yl)methanol
Formel: C₇H₆ClN₃O
Lagertemperatur: Raumtemperatur
CAS-Nummer: 1896916-30-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR