2-Bromoimidazo[1,2-a]pyridine-7-carboxylic acid ≥98%

Lieferant: Apollo Scientific

OR510176-100MG
APOSOR510176-100MGEA 353 EUR
APOSOR510176-100MG
2-Bromoimidazo[1,2-a]pyridine-7-carboxylic acid ≥98%
2-Bromoimidazo[1,2-a]pyridine-7-carboxylic acid
Formel: C₈H₅BrN₂O₂
Lagertemperatur: Raumtemperatur
CAS-Nummer: 1784089-67-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR