7-Bromo-5-methoxybenzo[d]thiazol-2-amine ≥98%

Lieferant: Apollo Scientific

OR510177-100MG
APOSOR510177-100MGEA 337 EUR
APOSOR510177-100MG
7-Bromo-5-methoxybenzo[d]thiazol-2-amine ≥98%
7-Bromo-5-methoxybenzo[d]thiazol-2-amine
Formel: C₈H₇BrN₂OS
Lagertemperatur: Raumtemperatur
MDL: MFCD31744450
CAS-Nummer: 1998062-49-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR