Methyl 5-bromo-4-methyl-1H-indole-2-carboxylate ≥98%

Lieferant: Apollo Scientific

OR510178-250MG
APOSOR510178-250MGEA 364 EUR
APOSOR510178-250MG
Methyl 5-bromo-4-methyl-1H-indole-2-carboxylate ≥98%
Methyl 5-bromo-4-methyl-1H-indole-2-carboxylate
Formel: C₁₁H₁₀BrNO₂
Molecular Weight: 268,107 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29035115
CAS-Nummer: 1857296-39-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR