4,5,6,7-Tetrahydro-1H-indazole-5-carboxylic acid ≥98%
Lieferant: Apollo Scientific
OR510180-100MG
APOSOR510180-100MGEA
140
EUR
APOSOR510180-100MG
4,5,6,7-Tetrahydro-1H-indazole-5-carboxylic acid ≥98%
4,5,6,7-Tetrahydro-1H-indazole-5-carboxylic acid
Formel:
C₈H₁₀N₂O₂ Molecular Weight: 166.177 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20647600 CAS-Nummer: 52834-38-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...