4-Acetyl-3-hydroxybenzoic acid ≥98%

Lieferant: Apollo Scientific

OR510183-100MG
APOSOR510183-100MGEA 154 EUR
APOSOR510183-100MG
4-Acetyl-3-hydroxybenzoic acid ≥98%
4-Acetyl-3-hydroxybenzoic acid
Formel: C₉H₈O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00854034
CAS-Nummer: 102297-62-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR