4-Hydroxy-5-methoxy-2-nitrobenzoic acid ≥98%

Lieferant: Apollo Scientific

OR510185-100MG
APOSOR510185-100MGEA 279 EUR
APOSOR510185-100MG
4-Hydroxy-5-methoxy-2-nitrobenzoic acid ≥98%
4-Hydroxy-5-methoxy-2-nitrobenzoic acid
Formel: C₈H₇NO₆
Molecular Weight: 213,144 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22627819
CAS-Nummer: 140647-01-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR