7-Bromopyrido[3,2-d]pyrimidine-2,4(1H,3H)-dione ≥98%
Lieferant: Apollo Scientific
Synonyme:
7-Bromopyrido[3,2-d]pyrimidine-2,4-diol
OR510186-100MG
APOSOR510186-100MGEA
42.2
EUR
APOSOR510186-100MG
7-Bromopyrido[3,2-d]pyrimidine-2,4(1H,3H)-dione ≥98%
7-Bromopyrido[3,2-d]pyrimidine-2,4(1H,3H)-dione
Formel:
C₇H₄BrN₃O₂ Molecular Weight: 242,03 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13250073 CAS-Nummer: 1215074-37-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...