6-Chloro-2-morpholinonicotinic acid ≥98%

Lieferant: Apollo Scientific

OR510187-100MG
APOSOR510187-100MGEA 763 EUR
APOSOR510187-100MG
6-Chloro-2-morpholinonicotinic acid ≥98%
6-Chloro-2-morpholinonicotinic acid
Formel: C₁₀H₁₁ClN₂O₃
Molecular Weight: 242,659 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00452940
CAS-Nummer: 305863-07-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR