5-Bromo-4-chloro-3-nitro-1H-pyrrolo[2,3-b]pyridine ≥98%

Lieferant: Apollo Scientific

OR510189-100MG
APOSOR510189-100MGEA 432 EUR
APOSOR510189-100MG
5-Bromo-4-chloro-3-nitro-1H-pyrrolo[2,3-b]pyridine ≥98%
5-Bromo-4-chloro-3-nitro-1H-pyrrolo[2,3-b]pyridine
Formel: C₇H₃BrClN₃O₂
Molecular Weight: 276,475 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977512
CAS-Nummer: 1477532-48-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR