Methyl 5-iodo-1H-indazole-6-carboxylate ≥98%

Lieferant: Apollo Scientific

OR510190-100MG
APOSOR510190-100MGEA 480 EUR
APOSOR510190-100MG
Methyl 5-iodo-1H-indazole-6-carboxylate ≥98%
Methyl 5-iodo-1H-indazole-6-carboxylate
Formel: C₉H₇IN₂O₂
Molecular Weight: 302,069 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 1227268-85-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR