3-Iodo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazole-4-carbaldehyde ≥98%

Lieferant: Apollo Scientific

OR510191-1G
APOSOR510191-1GEA 1010 EUR
APOSOR510191-1G
3-Iodo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazole-4-carbaldehyde ≥98%
3-Iodo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazole-4-carbaldehyde
Formel: C₉H₁₁IN₂O₂
Molecular Weight: 306.1 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 1627924-19-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR