4,6-Dichloro-3-iodo-1H-pyrazolo[3,4-d]pyrimidine 95%

Lieferant: Apollo Scientific

OR510192-1G
APOSOR510192-1GEA 89.9 EUR
APOSOR510192-1G
4,6-Dichloro-3-iodo-1H-pyrazolo[3,4-d]pyrimidine 95%
4,6-Dichloro-3-iodo-1H-pyrazolo[3,4-d]pyrimidine
Formel: C₅HCl₂IN₄
Molecular Weight: 314,899 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD20258005
CAS-Nummer: 1379308-33-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR