2-(Tetrahydro-2H-pyran-2-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-indazole ≥98%

Lieferant: Apollo Scientific

OR510193-1G
APOSOR510193-1GEA 829 EUR
APOSOR510193-1G
2-(Tetrahydro-2H-pyran-2-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-indazole ≥98%
2-(Tetrahydro-2H-pyran-2-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-indazole
Formel: C₁₈H₂₅BN₂O₃
Molecular Weight: 328.214 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22124621
CAS-Nummer: 1146955-35-2

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR