(S)-Azepan-2-ylmethanol ≥98%
Lieferant: Apollo Scientific
OR510194-500MG
APOSOR510194-500MGEA
453
EUR
APOSOR510194-500MG
(S)-Azepan-2-ylmethanol ≥98%
(S)-Azepan-2-ylmethanol
Formel:
C₇H₁₅NO Molecular Weight: 129,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19224940 CAS-Nummer: 1314999-26-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...