(2R)-1-Azabicyclo[2.2.2]octane-2-carbonitrile ≥98%
Lieferant: Apollo Scientific
OR510196-100MG
APOSOR510196-100MGEA
839
EUR
APOSOR510196-100MG
(2R)-1-Azabicyclo[2.2.2]octane-2-carbonitrile ≥98%
(2R)-1-Azabicyclo[2.2.2]octane-2-carbonitrile
Formel:
C₈H₁₂N₂ Molecular Weight: 136,194 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18374363 CAS-Nummer: 885517-05-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...