(S)-2-methylpiperidine-2-carboxylic acid ≥98%

Lieferant: Apollo Scientific

OR510197-1G
APOSOR510197-1GEA 1970 EUR
APOSOR510197-1G
(S)-2-methylpiperidine-2-carboxylic acid ≥98%
(S)-2-methylpiperidine-2-carboxylic acid
Formel: C₇H₁₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18207175
CAS-Nummer: 89115-95-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR