(1R,2R,4R)-rel-1-Azabicyclo[2,2,1]heptane-2-carboxylic acid hydrochloride ≥98%

Lieferant: Apollo Scientific

OR510198-1G
APOSOR510198-1GEA 1340 EUR
APOSOR510198-1G
(1R,2R,4R)-rel-1-Azabicyclo[2,2,1]heptane-2-carboxylic acid hydrochloride ≥98%
(1R,2R,4R)-rel-1-Azabicyclo[2,2,1]heptane-2-carboxylic acid hydrochloride
Formel: C₇H₁₁NO₂·ClH
Molecular Weight: 177,629 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22572789
CAS-Nummer: 921755-46-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR