2-(3,5-Dichloropyridin-4-yl)acetic acid ≥98%
Lieferant: Apollo Scientific
OR510199-1G
APOSOR510199-1GEA
1510
EUR
APOSOR510199-1G
2-(3,5-Dichloropyridin-4-yl)acetic acid ≥98%
2-(3,5-Dichloropyridin-4-yl)acetic acid
Formel:
C₇H₅Cl₂NO₂ Molecular Weight: 206,026 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09925365 CAS-Nummer: 227781-56-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...