1-(7-Chloro-1H-indol-3-yl)-N,N-dimethylmethanamine ≥98%

Lieferant: Apollo Scientific

OR510200-100MG
APOSOR510200-100MGEA 753 EUR
APOSOR510200-100MG
1-(7-Chloro-1H-indol-3-yl)-N,N-dimethylmethanamine ≥98%
1-(7-Chloro-1H-indol-3-yl)-N,N-dimethylmethanamine
Formel: C₁₁H₁₃ClN₂
Molecular Weight: 208,687 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 159503-76-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR