2-Chloro-1-(1H-pyrrolo[2,3-b]pyridin-3-yl)ethan-1-one hydrochloride ≥98%
Lieferant: Apollo Scientific
OR510203-1G
APOSOR510203-1GEA
2220
EUR
APOSOR510203-1G
2-Chloro-1-(1H-pyrrolo[2,3-b]pyridin-3-yl)ethan-1-one hydrochloride ≥98%
2-Chloro-1-(1H-pyrrolo[2,3-b]pyridin-3-yl)ethan-1-one hydrochloride
Formel:
C₉H₇ClN₂O·ClH Molecular Weight: 231,079 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12197050 CAS-Nummer: 1181457-88-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...