7-Bromo-5-chloro-1H-benzimidazol-2-amine ≥98%

Lieferant: Apollo Scientific

OR510204-1G
APOSOR510204-1GEA 1300 EUR
APOSOR510204-1G
7-Bromo-5-chloro-1H-benzimidazol-2-amine ≥98%
7-Bromo-5-chloro-1H-benzimidazol-2-amine
Formel: C₇H₅BrClN₃
Molecular Weight: 246,492 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 1388021-17-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR