3-Bromo-2,4,5-trimethylaniline ≥98%
Lieferant: Apollo Scientific
OR510206-100MG
APOSOR510206-100MGEA
554
EUR
APOSOR510206-100MG
3-Bromo-2,4,5-trimethylaniline ≥98%
3-Bromo-2,4,5-trimethylaniline
Formel:
C₉H₁₂BrN Molecular Weight: 214,102 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD24623730 CAS-Nummer: 18087-53-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...