2-Bromo-7-chlorobenzo[b]thiophene ≥98%

Lieferant: Apollo Scientific

OR510207-1G
APOSOR510207-1GEA 2070 EUR
APOSOR510207-1G
2-Bromo-7-chlorobenzo[b]thiophene ≥98%
2-Bromo-7-chlorobenzo[b]thiophene
Formel: C₈H₄BrClS
Lagertemperatur: Raumtemperatur
CAS-Nummer: 1782669-70-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR