5-Bromo-1,2-dichloro-3-nitrobenzene ≥98%

Lieferant: Apollo Scientific

OR510209-100MG
APOSOR510209-100MGEA 1220 EUR
APOSOR510209-100MG
5-Bromo-1,2-dichloro-3-nitrobenzene ≥98%
5-Bromo-1,2-dichloro-3-nitrobenzene
Formel: C₆H₂BrCl₂NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD28250219
CAS-Nummer: 1805596-72-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR