5-Bromo-8-chloro-2-hydroxyquinolin-4(1H)-one ≥98%

Lieferant: Apollo Scientific

OR510210-1G
APOSOR510210-1GEA 753 EUR
APOSOR510210-1G
5-Bromo-8-chloro-2-hydroxyquinolin-4(1H)-one ≥98%
5-Bromo-8-chloro-2-hydroxyquinolin-4(1H)-one
Formel: C₉H₅BrClNO₂
Molecular Weight: 274,498 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 1996052-61-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR