4-Hydroxy-7-methoxy-6-nitroquinoline-3-carbonitrile ≥98%
Lieferant: Apollo Scientific
OR510212-100MG
APOSOR510212-100MGEA
644
EUR
APOSOR510212-100MG
4-Hydroxy-7-methoxy-6-nitroquinoline-3-carbonitrile ≥98%
4-Hydroxy-7-methoxy-6-nitroquinoline-3-carbonitrile
Formel:
C₁₁H₇N₃O₄ Molecular Weight: 245,191 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13181118 CAS-Nummer: 492456-52-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...