4-Bromo-6-cyanobenzo[b]thiophene-2-carboxylic acid ≥98%
Lieferant: Apollo Scientific
OR510213-500MG
APOSOR510213-500MGEA
2500
EUR
APOSOR510213-500MG
4-Bromo-6-cyanobenzo[b]thiophene-2-carboxylic acid ≥98%
4-Bromo-6-cyanobenzo[b]thiophene-2-carboxylic acid
Formel:
C₁₀H₄BrNO₂S Molecular Weight: 282,113 g/mol Lagertemperatur: Raumtemperatur |
CAS-Nummer:
1415564-52-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...