8-Iodo-3-nitro-1-naphthoic acid ≥98%

Lieferant: Apollo Scientific

OR510215-1G
APOSOR510215-1GEA 3130 EUR
APOSOR510215-1G
8-Iodo-3-nitro-1-naphthoic acid ≥98%
8-Iodo-3-nitro-1-naphthoic acid
Formel: C₁₁H₆INO₄
Molecular Weight: 343,074 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 67522-37-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR