N3-Methylpyridine-2,3-diamine ≥98%

Lieferant: Apollo Scientific

OR510217-100MG
APOSOR510217-100MGEA 78.3 EUR
APOSOR510217-100MG
N3-Methylpyridine-2,3-diamine ≥98%
N3-Methylpyridine-2,3-diamine
Formel: C₆H₉N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00490112
CAS-Nummer: 56291-51-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR