1-Oxa-7-azaspiro[4.5]decane hydrochloride ≥98%

Lieferant: Apollo Scientific

OR510219-100MG
APOSOR510219-100MGEA 202 EUR
APOSOR510219-100MG
1-Oxa-7-azaspiro[4.5]decane hydrochloride ≥98%
1-Oxa-7-azaspiro[4.5]decane hydrochloride
Formel: C₈H₁₅NO·ClH
Molecular Weight: 177,672 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22572788
CAS-Nummer: 1415562-85-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR