N4-[4-[4-(N-[4-[2-(1-Naphthyl)anilino]phenyl]anilino)phenyl]phenyl]-N1-[2-(1-naphthyl)phenyl]-N4-phenyl-benzene-1,4-diamine 99%

Lieferant: Apollo Scientific

Synonyme: N1,N1'-([1,1'-Biphenyl]-4,4'-diyl)bis(N4-(naphthalen-1-yl)-N1,N4-diphenylbenzene-1,4-diamine)

OR51022-5G
APOSOR51022-5GEA 66.7 EUR
APOSOR51022-5G
N4-[4-[4-(N-[4-[2-(1-Naphthyl)anilino]phenyl]anilino)phenyl]phenyl]-N1-[2-(1-naphthyl)phenyl]-N4-phenyl-benzene-1,4-diamine 99%
N4-[4-[4-(N-[4-[2-(1-Naphthyl)anilino]phenyl]anilino)phenyl]phenyl]-N1-[2-(1-naphthyl)phenyl]-N4-phenyl-benzene-1,4-diamine
Formel: C₆₈H₅₀N₄
Molecular Weight: 923.151 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135499
CAS-Nummer: 209980-47-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR