7-Chlorochroman-4-amine ≥98%

Lieferant: Apollo Scientific

OR510220-100MG
APOSOR510220-100MGEA 0 EUR
APOSOR510220-100MG
7-Chlorochroman-4-amine ≥98%
7-Chlorochroman-4-amine
Formel: C₉H₁₀ClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD17010010
CAS-Nummer: 763907-56-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR