(R)-5,6,7,8-tetrahydroquinolin-8-amine hydrochloride ≥98%
Lieferant: Apollo Scientific
OR510221-100MG
APOSOR510221-100MGEA
85.7
EUR
APOSOR510221-100MG
(R)-5,6,7,8-tetrahydroquinolin-8-amine hydrochloride ≥98%
(R)-5,6,7,8-tetrahydroquinolin-8-amine hydrochloride
Formel:
C₉H₁₂N₂·ClH Molecular Weight: 184,666 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31654063 CAS-Nummer: 1431726-92-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...