4-Hydroxy-2-naphthoic acid ≥98%
Lieferant: Apollo Scientific
OR510222-100MG
APOSOR510222-100MGEA
256
EUR
APOSOR510222-100MG
4-Hydroxy-2-naphthoic acid ≥98%
4-Hydroxy-2-naphthoic acid
Formel:
C₁₁H₈O₃ Molecular Weight: 188,179 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12405016 CAS-Nummer: 1573-91-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...