1-Benzyl-6-oxa-1-azaspiro[3.3]heptane ≥98%

Lieferant: Apollo Scientific

OR510223-100MG
APOSOR510223-100MGEA 322 EUR
APOSOR510223-100MG
1-Benzyl-6-oxa-1-azaspiro[3.3]heptane ≥98%
1-Benzyl-6-oxa-1-azaspiro[3.3]heptane
Formel: C₁₂H₁₅NO
Molecular Weight: 189,254 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19443155
CAS-Nummer: 1223573-38-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR