2-Bromo-3-methoxy-2-cyclopenten-1-one ≥98%
Lieferant: Apollo Scientific
OR510224-100MG
APOSOR510224-100MGEA
230
EUR
APOSOR510224-100MG
2-Bromo-3-methoxy-2-cyclopenten-1-one ≥98%
2-Bromo-3-methoxy-2-cyclopenten-1-one
Formel:
C₆H₇BrO₂ Molecular Weight: 191.023 g/mol Lagertemperatur: Raumtemperatur |
CAS-Nummer:
14203-25-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...