4-Amino-2-chloro-3-methylbenzonitrile hydrochloride ≥98%

Lieferant: Apollo Scientific

OR510226-250MG
APOSOR510226-250MGEA 88.6 EUR
APOSOR510226-250MG
4-Amino-2-chloro-3-methylbenzonitrile hydrochloride ≥98%
4-Amino-2-chloro-3-methylbenzonitrile hydrochloride
Formel: C₈H₇ClN₂·ClH
Molecular Weight: 203,069 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11858333
CAS-Nummer: 864361-78-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR