7-Chloro-3-methylquinoline-8-carboxylic acid ≥98%

Lieferant: Apollo Scientific

OR510227-5G
APOSOR510227-5GEA 227 EUR
APOSOR510227-5G
7-Chloro-3-methylquinoline-8-carboxylic acid ≥98%
7-Chloro-3-methylquinoline-8-carboxylic acid
Formel: C₁₁H₈ClNO₂
Molecular Weight: 221,64 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00145199
CAS-Nummer: 90717-03-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR