7-Bromo-2-methyl-8-quinolinamine ≥98%

Lieferant: Apollo Scientific

OR510229-100MG
APOSOR510229-100MGEA 404 EUR
APOSOR510229-100MG
7-Bromo-2-methyl-8-quinolinamine ≥98%
7-Bromo-2-methyl-8-quinolinamine
Formel: C₁₀H₉BrN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD12402358
CAS-Nummer: 1216061-60-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR