2,6-Dibromo-9,10-bis(4-tert-butylphenyl)anthracene 99%

Lieferant: Apollo Scientific

OR51023-1G
APOSOR51023-1GEA 438 EUR
APOSOR51023-1G
2,6-Dibromo-9,10-bis(4-tert-butylphenyl)anthracene 99%
2,6-Dibromo-9,10-bis(4-tert-butylphenyl)anthracene
Formel: C₃₄H₃₂Br₂
Molecular Weight: 600,44 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 1187763-68-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR