6-Bromo-8-aminoquinaldine ≥98%

Lieferant: Apollo Scientific

OR510230-100MG
APOSOR510230-100MGEA 404 EUR
APOSOR510230-100MG
6-Bromo-8-aminoquinaldine ≥98%
6-Bromo-8-aminoquinaldine
Formel: C₁₀H₉BrN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD22571456
CAS-Nummer: 61854-65-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR