1-(7-Bromobenzo[d][1,3]dioxol-4-yl)ethan-1-one ≥98%

Lieferant: Apollo Scientific

OR510231-1G
APOSOR510231-1GEA 112 EUR
APOSOR510231-1G
1-(7-Bromobenzo[d][1,3]dioxol-4-yl)ethan-1-one ≥98%
1-(7-Bromobenzo[d][1,3]dioxol-4-yl)ethan-1-one
Formel: C₉H₇BrO₃
Molecular Weight: 243,054 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30738025
CAS-Nummer: 1892297-27-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR