Methyl 3-bromo-4-methoxy-2-methylbenzoate ≥98%
Lieferant: Apollo Scientific
OR510232-250MG
APOSOR510232-250MGEA
106
EUR
APOSOR510232-250MG
Methyl 3-bromo-4-methoxy-2-methylbenzoate ≥98%
Methyl 3-bromo-4-methoxy-2-methylbenzoate
Formel:
C₁₀H₁₁BrO₃ Molecular Weight: 259,097 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23703479 CAS-Nummer: 1427501-73-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...